Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

450294

Sigma-Aldrich

Sodium selenate decahydrate

99.999% trace metals basis

Synonym(s):

Disodium selenate decahydrate, Sodium selenate (Na2 SeO4 ) decahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2SeO4 · 10H2O
CAS Number:
Molecular Weight:
369.09
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

99.999% trace metals basis

form

crystalline

mp

35 °C (dec.) (lit.)

density

1.6 g/mL at 25 °C

SMILES string

[Na+].[Na+].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[O-][Se]([O-])(=O)=O

InChI

1S/2Na.H2O4Se.10H2O/c;;1-5(2,3)4;;;;;;;;;;/h;;(H2,1,2,3,4);10*1H2/q2*+1;;;;;;;;;;;/p-2

InChI key

TYVNFQRMDPFYPC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

signalword

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Álvarez-Fernández García et al.
The Analyst, 145(4), 1457-1465 (2019-12-24)
Several organisms have demonstrated the ability of synthesising biogenic selenium-containing nanoparticles. Such particles from biological sources have attracted great attention due to several proven activities as antioxidants or antimicrobial agents. However, little is known in terms of size (distribution), shapes
W C Stewart et al.
Journal of animal science, 90(2), 577-584 (2011-10-04)
Adequate Se transfer from ewes to lambs is important to prevent Se-deficiency diseases. To evaluate how different chemical forms of Se administered at comparative dosages to mature ewes affect Se status of their lambs, 240 ewes were divided into 8
Beatrice Pezzarossa et al.
Journal of the science of food and agriculture, 92(4), 781-786 (2011-09-29)
We investigated the effects of the foliar and fruit application of sodium selenate on selenium (Se) accumulation, fruit growth and ripening in peach and pear. Trials were conducted in two growing seasons. In 2008 selenate was applied at a rate
N A Golubkina et al.
Doklady biological sciences : proceedings of the Academy of Sciences of the USSR, Biological sciences sections, 444, 176-179 (2012-07-05)
Accumulation and translocation intensity of selenium nanoparticles by natural selenium accumulators-perennial onions-is established and a decrease of these parameters is demonstrated in a sequence: Se(+6) > Se(0) > Se(+4).
N M Corcoran et al.
British journal of cancer, 103(4), 462-468 (2010-07-22)
Angiogenesis is fundamental to the progression of many solid tumours including prostate cancer. Sodium selenate is a small, water-soluble, orally bioavailable activator of PP2A phosphatase with anti-angiogenic properties. This was a dose-escalation phase I study in men with asymptomatic, chemotherapy-naïve

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service