Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

446432

Sigma-Aldrich

7-tert-Butoxy-2,5-norbornadiene

96%

Synonym(s):

7-(Tert-butoxy)norbornadiene, 7-tert -Butoxy-2,5-norbornadiene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H16O
CAS Number:
Molecular Weight:
164.24
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

96%

refractive index

n20/D 1.469 (lit.)

bp

70-72 °C/14 mmHg (lit.)

density

0.936 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)OC1C2C=CC1C=C2

InChI

1S/C11H16O/c1-11(2,3)12-10-8-4-5-9(10)7-6-8/h4-10H,1-3H3/t8-,9+,10?

InChI key

PTTSCAGWOHYHDN-ULKQDVFKSA-N

pictograms

Flame

signalword

Warning

hcodes

Hazard Classifications

Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

127.4 °F - closed cup

flash_point_c

53 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service