Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

44506

Sigma-Aldrich

Aliquat® HTA-1

High-Temperature Phase Transfer Catalyst, 30-35% in H2O

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352000

concentration

30-35% in H2O

pH

6-9

density

1.055 g/mL at 20 °C (lit.)

General description

Virtually all quaternary ammonium based PTC catalysts decompose below 100 °C and/or in the presence of caustics. Aliquat® HTA-1 has been found to be stable at temperatures well above 100 °C

Legal Information

Aliquat is a registered trademark of BASF

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service