Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

444480

Sigma-Aldrich

Polyethylene monoalcohol

average Mn ~700, crystalline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2CH2)nCH2OH
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

crystalline

mol wt

average Mn ~700

impurities

15-20% unreacted hydrocarbons

viscosity

7.9 cP(149 °C, Brookfield)(lit.)

hydroxyl value

65 mg KOH/g

transition temp

Tm 108 °C

solubility

toluene, MEK and hexane: soluble

density

0.985 g/mL at 25 °C (lit.)

Mw/Mn

~1.09

SMILES string

C=C

InChI

1S/C14H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h15H,2-14H2,1H3

InChI key

HLZKNKRTKFSKGZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

Precursor to mono-carboxylic acids, -sulfates and -amines

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aurelio Ramírez-Hernández et al.
Carbohydrate research, 488, 107907-107907 (2020-01-24)
The objective of this paper was to perform a copolymerization between polyethylene and starch in order to obtain new environmentally friendly materials. The copolymer obtained was characterized thermally, morphologically and structurally, including its pasting profile. The starch-g-PE copolymer showed lower

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service