Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

444146

Sigma-Aldrich

Scandium(III) perchlorate solution

40 wt. % in H2O

Synonym(s):

Scandium perchlorate, Scandium triperchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sc(ClO4)3
CAS Number:
Molecular Weight:
361.32
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
form:
liquid

vapor pressure

0.12 psi ( 20 °C)
1.68 psi ( 55 °C)

form

liquid

reaction suitability

reagent type: oxidant

concentration

38-40% (EDTA titration)
40 wt. % in H2O

density

1.449 g/mL at 25 °C

SMILES string

[Sc+3].[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O

InChI

1S/3ClHO4.Sc/c3*2-1(3,4)5;/h3*(H,2,3,4,5);/q;;;+3/p-3

Inchi Key

UBVKRMCXDCYZQK-UHFFFAOYSA-K

Application

Excellent and reusable catalyst for α-C- and N-glycosylation reactions.[1]

Features and Benefits

Excellent and reusable catalyst for α-C- and N-glycosylation reactions. Hachiya, I. Kobayashi, S. Tetrahedron Lett. 1994, 35, 3319.

Pictograms

Flame over circleCorrosion

signalword

Danger

hcodes

Hazard Classifications

Ox. Liq. 2 - Skin Corr. 1B

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 35, 3319-3319 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service