Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

443832

Sigma-Aldrich

Hexacontane

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)58CH3
CAS Number:
Molecular Weight:
843.61
Beilstein/REAXYS Number:
1810088
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

assay

98%

form

powder

mp

96-101 °C

SMILES string

CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

InChI

1S/C60H122/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35-37-39-41-43-45-47-49-51-53-55-57-59-60-58-56-54-52-50-48-46-44-42-40-38-36-34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h3-60H2,1-2H3

InChI key

OCWUCHKZAHTZAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct separation and quantitative determination of (n-, iso-) alkanes in neat asphalt using urea adduction and high-temperature gas chromatography (HTGC).
Netzel DA and Rovani JF.
Energy and Fuels, 21(1), 333-338 (2007)
Small-Angle X-ray Scattering of n-Hexacontane as a Function of Temperature.
Mandelkern L, et al.
Macromolecules, 27(8), 2324-2326 (1994)
Wei Xu et al.
Molecules (Basel, Switzerland), 17(12), 14323-14335 (2012-12-05)
An ionic liquids (IL)-based microwave-assisted approach for extraction and determination of flavonoids from Bauhinia championii (Benth.) Benth. was proposed for the first time. Several ILs with different cations and anions and the microwave-assisted extraction (MAE) conditions, including sample particle size
Juxia Fu et al.
The journal of physical chemistry. A, 109(51), 11724-11732 (2005-12-22)
The nature of the linear dichroism in the near-edge X-ray absorption fine structure (NEXAFS) spectra of linear n-alkanes is a matter of long-standing controversy. Linear dichroism in the carbon 1s --> sigma*(C-C) transition has been interpreted within a building block
Erich A Müller et al.
The journal of physical chemistry. B, 115(44), 12822-12834 (2011-09-22)
Canonical ensemble molecular dynamics (MD) simulations are reported which compute both the vapor-liquid equilibrium properties (vapor pressure and liquid and vapor densities) and the interfacial properties (density profiles, interfacial tensions, entropy and enthalpy of surface formation) of four long-chained n-alkanes:

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service