Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

443697

Sigma-Aldrich

Ethyl 3-amino-4,4,4-trifluorocrotonate

96%

Synonym(s):

Ethyl 3-amino-4,4,4-trifluoro-2-butenoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C(NH2)=CHCO2C2H5
CAS Number:
Molecular Weight:
183.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

96%

form

liquid

refractive index

n20/D 1.424 (lit.)

bp

83 °C/15 mmHg (lit.)

density

1.245 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)C=C(N)C(F)(F)F

InChI

1S/C6H8F3NO2/c1-2-12-5(11)3-4(10)6(7,8)9/h3H,2,10H2,1H3

InChI key

NXVKRKUGIINGHD-UHFFFAOYSA-N

General description

Ethyl 3-amino-4,4,4-trifluorocrotonate is an ester.

Application

Ethyl 3-amino-4,4,4-trifluorocrotonate may be employed for the preparation of 4-trifluoromethyl-1,3-oxazin-6-ones[1] and 4-quinolinone derivatives.[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

149.0 °F - closed cup

flash_point_c

65 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Heterocyclic Chemistry, 30, 71-71 (1993)
Aliphatic fluorine-containing amino acids.
Kukhar VP and Soloshonok VA.
Russ. Chem. Rev., 60(8), 850-850 (1991)
Journal of Heterocyclic Chemistry, 30, 49-49 (1993)
Syntheses of New 4-Trifluoromethylated 1, 3-Oxazin-6-Ones from the Enamine of Ethyl Trifluoroacetoacetate.
Decock-Plancquaert M-a, et al.
Bull. Soc. Chim. Belg., 101(4), 313-321 (1992)
A convenient route to diverse heterocycles through an addition of ?-amino carbonyl compounds to 3-halogeno-4-methoxybenzynes.
Yoon K, et al.
The Journal of Organic Chemistry, 70(14), 5741-5744 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service