Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

442402

Sigma-Aldrich

Poly(styrene-co-maleic anhydride), cumene terminated

average Mn ~1,900 by GPC

Synonym(s):

Poly(styrene-co-maleic anhydride), maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

mol wt

average Mn ~1,900 by GPC

composition

styrene, 75 wt. %

acid number

265‑305 mg KOH/g

transition temp

Tg 125 °C

solubility

ketones, ethers and alkali: soluble

InChI

1S/C9H12.C8H8.C4H2O3/c1-8(2)9-6-4-3-5-7-9;1-2-8-6-4-3-5-7-8;5-3-1-2-4(6)7-3/h3-8H,1-2H3;2-7H,1H2;1-2H

InChI key

QQNZOECCMADQHL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Leveling aids in films, embrittling/anti-resoil agents in shampoos, pigment dispersants, paper sizing and aqueous grinding fluids.

Features and Benefits

Low solution viscosity and high functionality.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service