Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

441945

Sigma-Aldrich

(Hydroxymethyl)benzoguanamine, methylated/ethylated

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

description

130 - 190 wt. per reaction with hydroxyl, carboxyl or amide

assay

≥98%

form

liquid

refractive index

n20/D 1.559 (lit.)

Gardner-Holdt

27.0-63.4 stokes(25 °C)(lit.)

density

1.164 g/mL at 25 °C (lit.)

Application

(Hydroxymethyl)benzoguanamine ethylated may be used:
  • As a cross-linking reagent.[1]
  • In the synthesis of nonlinear optically active (NLO) chromophores based on amino azobenzene dyes.[2]
Contributes good chemical, detergent and salt spray resistance to films.
Cross-linking agent for electrocoating and conventional coatings requiring high detergent and chemical resistance.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Skin Sens. 1

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New Antireflective Coating Materials Containing a Novel Chromophore for KrF Laser Lithography.
Ahn K-D, et al.
J. Photopolym. Sci. Technol., 14(3), 475-480 (2001)
Preparation and characterization of all organic NLO sol-gel materials based on amino azobenzene dyes.
Hsiue G-H, et al.
Macromolecular Chemistry and Physics, 201(17), 2336-2347 (2000)
A low-temperature-cross-linked poly (4-vinylphenol) gate-dielectric for organic thin film transistors.
Vicca P, et al.
Thin Solid Films, 519(1), 391-393 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service