Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

441031

Sigma-Aldrich

(1S)-(+)-Menthyl acetate

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O2
CAS Number:
Molecular Weight:
198.30
Beilstein/REAXYS Number:
1909419
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:

assay

99%

form

liquid

optical activity

[α]20/D +80°, c = 8 in benzene

optical purity

ee: 98% (GLC)

refractive index

n20/D 1.446 (lit.)

bp

228-229 °C (lit.)

density

0.925 g/mL at 25 °C (lit.)

SMILES string

CC(C)[C@H]1CC[C@H](C)C[C@@H]1OC(C)=O

InChI

1S/C12H22O2/c1-8(2)11-6-5-9(3)7-12(11)14-10(4)13/h8-9,11-12H,5-7H2,1-4H3/t9-,11+,12-/m0/s1

InChI key

XHXUANMFYXWVNG-WCQGTBRESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

170.6 °F - closed cup

flash_point_c

77 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service