Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

440892

Sigma-Aldrich

Nornicotine

97%

Synonym(s):

3-(2-Pyrrolidinyl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12N2
CAS Number:
Molecular Weight:
148.20
UNSPSC Code:
12352005
PubChem Substance ID:
Pricing and availability is not currently available.

assay

97%

InChI

1S/C9H12N2/c1-3-8(7-10-5-1)9-4-2-6-11-9/h1,3,5,7,9,11H,2,4,6H2

InChI key

MYKUKUCHPMASKF-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fatemeh Rajabi et al.
PloS one, 12(1), e0169778-e0169778 (2017-01-13)
The great potential of pharmacologically active secondary plant metabolites is often limited by low yield and availability of the producing plant. Chemical synthesis of these complex compounds is often too expensive. Plant cell fermentation offers an alternative strategy to overcome
Ramsey S Lewis et al.
Frontiers in plant science, 11, 368-368 (2020-04-23)
Genetic methodologies for reducing nicotine accumulation in the tobacco plant (Nicotiana tabacum L.) are of interest because of potential future regulations that could mandate lowering of this alkaloid in conventional cigarettes. Inactivation of tobacco genes such as the Berberine Bridge
Huimin Deng et al.
Journal of separation science, 40(23), 4645-4652 (2017-09-30)
Nornicotine, an alkaloid constituent of tobacco, is a precursor to the carcinogen N-nitrosonornicotine that is produced during the curing and processing of tobacco. Accumulating evidence reveals that nornicotine enantiomers have different neurochemical and behavioral effects. In the present study, an
Andrew C Harris et al.
Drug and alcohol dependence, 185, 1-9 (2018-02-08)
Animal models are needed to inform FDA regulation of electronic cigarettes (ECs) because they avoid limitations associated with human studies. We previously reported that an EC refill liquid produced less aversive/anhedonic effects at a high nicotine dose than nicotine alone
Yufei Wang et al.
Psychopharmacology, 237(9), 2809-2822 (2020-06-20)
The behavioral effects of the nicotine metabolites nornicotine and cotinine have not been investigated extensively. To evaluate the effects of nicotine, cotinine, and nornicotine, given alone or in combination, on locomotor activity and emission of ultrasonic vocalizations in male adult

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service