Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

440043

Sigma-Aldrich

Poly(4-methyl-1-pentene)

low molecular weight

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH[CH2CH(CH3)2])n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

beads

melt index

180 g/min. (260°C/5kg)

dielectric constant

2.12, 100 Hz (ASTM D 150)

hardness

80 (Rockwell R, ASTM D 785)

mp

240 °C

transition temp

softening point 173 °C (Vicat, ASTM D 1525)

density

0.833 g/mL at 25 °C

SMILES string

CC(C)CC=C

InChI

1S/C6H12/c1-4-5-6(2)3/h4,6H,1,5H2,2-3H3

InChI key

WSSSPWUEQFSQQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Coating for paper food containers for microwave and conventional ovens. Release coatings for food and synthetic leather. Molded into medical labware. Film for decorative laminates and printed circuit boards.

Features and Benefits

Very low specific gravity, high transparency, resistant to oxidation and prone to stress cracking. Good electrical insulator.

Physical form

Low density thermoplastic.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LaNetra M Clayton et al.
Journal of nanoscience and nanotechnology, 6(8), 2520-2524 (2006-10-14)
Poly(4-methyl-1-pentene), PMP, a high melting polymer composed of hydrogen and carbon, has the potential to become an alternative to polyethylene (PE) as shielding material against Galactic Cosmic Radiation (GCR). PMP exhibits higher thermal stability than PE and is transparent in
C Prasser et al.
European journal of anaesthesiology, 25(2), 152-157 (2007-07-28)
It has been demonstrated that volatile anaesthetics have cardioprotective properties during open-heart procedures, especially when administered continuously. European Council Directive 93/42/EEC concerning medical devices bans the supplementary incorporation of anaesthetic vaporizers in the bypass circuit. Since the uptake of volatile
Sang-Chul Shin et al.
International journal of pharmaceutics, 235(1-2), 141-147 (2002-03-07)
Triprolidine-containing matrix was fabricated with poly(4-methyl-1-pentene) (TPX) polymer to control the release of the drug. Effect of penetration enhancer and stripping of skin on the permeation of triprolidine through the excised mouse skin was studied. Penetrating enhancers showed the increased
Ayano Chiba et al.
Physical review. E, Statistical, nonlinear, and soft matter physics, 85(2 Pt 1), 021807-021807 (2012-04-03)
High-pressure in situ x-ray diffraction and specific-volume measurements on isotactic poly(4-methyl-1-pentene) melt have uncovered abrupt changes in the pressure dependence of microscopic structure as well as that of macroscopic density. The first sharp diffraction peak of the polymer melt, which
Michael T Demko et al.
ACS nano, 6(8), 6890-6896 (2012-07-18)
Soft lithography methods are emerging as useful tools for high-resolution, three-dimensional patterning of polymers and nanoparticles. However, the low Young's modulus of the standard template material, poly(dimethylsiloxane) (PDMS), limits attainable resolution, fidelity, and alignment capability. While much research has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service