Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

438790

Sigma-Aldrich

Yttrium(III) acetylacetonate hydrate

99.95% trace metals basis

Synonym(s):

Y(acac)3 hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y(C5H7O2)3 · xH2O
CAS Number:
Molecular Weight:
386.23 (anhydrous basis)
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

99.95% trace metals basis

form

powder or crystals

reaction suitability

core: yttrium
reagent type: catalyst

SMILES string

O.CC(=O)\C=C(\C)O[Y](O\C(C)=C/C(C)=O)O\C(C)=C/C(C)=O

InChI

1S/3C5H8O2.H2O.Y/c3*1-4(6)3-5(2)7;;/h3*3,6H,1-2H3;1H2;/q;;;;+3/p-3/b3*4-3-;;

InChI key

KGNBFQNJCULYHV-KJVLTGTBSA-K

General description

Atomic number of base material: 39 Yttrium

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Continuous efficiency improvements in photovoltaic devices result from material advancements and manufacturing innovation.

High Purity Metalorganic Precursors for CPV Device Fabrication

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service