Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

438057

Sigma-Aldrich

(S)-(+)-α-Aminocyclohexanepropionic acid hydrate

technical grade

Synonym(s):

3-Cyclohexyl-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2CH(NH2)CO2H · xH2O
CAS Number:
Molecular Weight:
171.24 (anhydrous basis)
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

technical grade

form

solid

optical activity

[α]21/D +12.0°, c = 1 in 1 M NaOH

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

mp

234-237 °C (lit.)

application(s)

peptide synthesis

SMILES string

[H]O[H].N[C@@H](CC1CCCCC1)C(O)=O

InChI

1S/C9H17NO2.H2O/c10-8(9(11)12)6-7-4-2-1-3-5-7;/h7-8H,1-6,10H2,(H,11,12);1H2/t8-;/m0./s1

InChI key

HDJQIWAIDCEDEF-QRPNPIFTSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service