Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

438022

Sigma-Aldrich

1-(Methoxymethyl)-1H-benzotriazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9N3O
CAS Number:
Molecular Weight:
163.18
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

99%

form

solid

mp

42-43 °C (lit.)

functional group

ether

SMILES string

COCn1nnc2ccccc12

InChI

1S/C8H9N3O/c1-12-6-11-8-5-3-2-4-7(8)9-10-11/h2-5H,6H2,1H3

InChI key

QEIXBXXKTUNWDK-UHFFFAOYSA-N

Application

1-(Methoxymethyl)-1H-benzotriazole can be used:
  • As a model compound in the study of photoelectron spectra of benzotriazoles, which can help in understanding their chemical reactivity.[1]
  • To prepare a rhenium(IV) complex named [HMEBTA]2[ReIVBr6] having magnetic properties.[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Self-assembly of the hexabromorhenate (IV) anion with protonated benzotriazoles: X-ray structure and magnetic properties
Martinez-Lillo J, et al.
Crystal Growth & Design, 14(11), 5985-5990 (2014)
Structural chemistry of polycyclic heteroaromatic compounds. Part 10: photoelectron spectra and electronic structures of 1-substituted 1H-benzotriazoles
Rademacher P, et al.
Journal of Molecular Structure, 513(1-3), 47-62 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service