437999
Sulfuryl chloride fluoride
technical grade, ≥90%
Synonym(s):
Chlorosulfonyl fluoride, Fluorosulfonyl chloride, Sulfuryl chlorofluoride
About This Item
Recommended Products
grade
technical grade
Quality Level
assay
≥90%
storage temp.
−20°C
SMILES string
FS(Cl)(=O)=O
InChI
1S/ClFO2S/c1-5(2,3)4
InChI key
IXPAAHZTOUOJJM-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Legal Information
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B
supp_hazards
Storage Class
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
wgk_germany
WGK 3
flash_point_f
235.4 °F - closed cup
flash_point_c
113 °C - closed cup
ppe
Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service