Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

437700

Sigma-Aldrich

Pyridine-d5

"100%", 99.96 atom % D, contains 0.03 % (v/v) TMS

Synonym(s):

Pentadeuteropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5D5N
CAS Number:
Molecular Weight:
84.13
Beilstein/REAXYS Number:
114377
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:

isotopic purity

99.96 atom % D

assay

99% (CP)

form

liquid

contains

0.03 % (v/v) TMS

expl. lim.

0.34-6.3 % (lit.)

technique(s)

NMR: suitable

refractive index

n20/D 1.506 (lit.)

pH

8.5 (0.2 g/L)

bp

114.4 °C (lit.)

mp

-41 °C (lit.)

density

1.05 g/mL at 25 °C (lit.)

mass shift

M+5

SMILES string

[2H]c1nc([2H])c([2H])c([2H])c1[2H]

InChI

1S/C5H5N/c1-2-4-6-5-3-1/h1-5H/i1D,2D,3D,4D,5D

InChI key

JUJWROOIHBZHMG-RALIUCGRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pyridine-d5 is a deuterated NMR solvent containing 0.03% (v/v) TMS (tetramethylsilane). It is useful in NMR-based research and analyses. It has been synthesized by palladium catalyzed H/D (hydrogen/deuterium) exchange reaction between pyridine vapor and heavy water. Infrared and Raman spectra of pyridine-d5 have been recorded in the range 300–4000cm-1.

Application

Pyridine-d5 may be used as a solvent in the 1H NMR based structural analysis of lignin acetates obtained from spruce and birch.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 2

flash_point_f

62.6 °F

flash_point_c

17 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NMR studies of lignins. 8. Examination of pyridine-d5 solutions of acetylated lignins from birch and spruce by 1H NMR spectroscopy.
Lundquist K and Von Unge S.
Acta Chemica Scandinavica. Series B, 40, 791-797 (1986)
Shao-Dan Chen et al.
Fitoterapia, 102, 67-73 (2015-02-24)
Five new diarylheptanoids (1-5), along with nine known ones (6-14), were isolated from the rhizomes of Curcuma kwangsiensis. Their structures were established on the basis of spectroscopic analyses. Compounds 1-3 were cyclic diarylheptanoids rarely discovered from C. kwangsiensis. Of all
Indra Prakash et al.
Natural product communications, 9(8), 1135-1138 (2014-09-23)
We report the isolation and complete structure of an isomer of rebaudioside D, known as rebaudioside D2. This novel steviol glycoside was isolated from a bioconversion reaction of rebaudioside A to rebaudioside D. Rebaudioside D2 possesses a relatively rare 1
Yu Shan et al.
Molecules (Basel, Switzerland), 20(11), 20334-20340 (2015-11-17)
A new nortriterpene saponin, 3-O-β-d-glucuronopyranosyl-30-norolean-12,20(29)-dien-23- oxo-28-oic acid, namely bigelovii D (11), was isolated from the hydroalcoholic extract of herbs of Salicornia bigelovii along with 10 known saponins (1-10). Their chemical structures were identified on the basis of spectroscopic analyses including
Xu Pang et al.
Carbohydrate research, 402, 236-240 (2014-12-17)
Five new steroidal saponins (1-5) were isolated from the fermentation broth of total furostanol glycosides from tubers of Dioscorea zingiberensis C.H. Wright incubated with a fungal, Absidia coerulea AS 3.3389, along with known saponins, zingiberensis new saponin (6), deltonin (7)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service