Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

437492

Sigma-Aldrich

Dodecamethylcyclohexasilane

≥97% (GC)

Synonym(s):

Dodecamethylcyclohexasilane, Dodecamethylcyclohexasilylene, Dodecamethylhexasilacyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(Si(CH3)2)6
CAS Number:
Molecular Weight:
348.93
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

≥97% (GC)

form

powder or crystals

mp

229 °C (lit.)

SMILES string

C[Si]1(C)[Si](C)(C)[Si](C)(C)[Si](C)(C)[Si](C)(C)[Si]1(C)C

InChI

1S/C12H36Si6/c1-13(2)14(3,4)16(7,8)18(11,12)17(9,10)15(13,5)6/h1-12H3

InChI key

RTCLHEHPUHREBC-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

atomic layer deposition (ALD), microelectronics, Mo:Al2O3 films, nanocomposite coating, photovoltaics, semiconductor devices, W:Al2O3 films, composite films, layer-by-layer

Silica's versatility spans various industries, including biomedical applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service