Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

437425

Sigma-Aldrich

Isooctyl acrylate

contains 75-125 ppm monomethyl ether hydroquinone as inhibitor, >90%

Synonym(s):

Isooctyl acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2(CH2)5CH(CH3)2
CAS Number:
Molecular Weight:
184.28
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

assay

>90%

form

liquid

contains

75-125 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.437 (lit.)

bp

125 °C/20 mmHg (lit.)

density

0.88 g/mL at 25 °C (lit.)

SMILES string

CC(C)CCCCCOC(=O)C=C

InChI

1S/C11H20O2/c1-4-11(12)13-9-7-5-6-8-10(2)3/h4,10H,1,5-9H2,2-3H3

InChI key

DXPPIEDUBFUSEZ-UHFFFAOYSA-N

General description

The structure of Isooctyl acrylate consists of eight carbon alkyl esters of acrylic acid. [1]

Application

Isooctyl acrylate may be used to prepare linear copolymers with cinnamoyloxyethyl methacrylate (CEMA) for applications as oil absorbents.[2] Copolymer of isooctyl acrylate /acrylamine (1:1 mole ratio) was studied as a magnetoelastic CO2 sensor With high sensitivity. [3] Isooctyl acrylate (IOA) based copolymers may be used to prepare pressure-sensitive acrylic medical grade adhesives for skin. [4][5]

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 2

flash_point_f

195.1 - 196.0 °F - Pensky-Martens closed cup

flash_point_c

90.6 - 91.1 °C - Pensky-Martens closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combinatorial Development of Pressure?Sensitive Adhesives.
Grunlan JC, et al.
Macromolecular Rapid Communications, 25(1), 286-291 (2004)
Q Y Cai et al.
Sensors and actuators. B, Chemical, 77(3), 614-619 (2002-08-27)
This paper presents a wireless, remote query ammonia sensor comprised of a free-standing magnetoelastic thick-film coated with a polymer, poly(acrylic acid-co-isooctylacrylate), that changes mass in response to atmospheric ammonia concentration. The mass of the polymer layer modulates the resonant frequency
A wireless, remote query magnetoelastic CO2 sensorElectronic Supplementary information available.
Cai QY, et al.
Journal of Environmental Monitoring, 2(6), 556-560 (2000)
Q Y Cai et al.
Sensors and actuators. B, Chemical, 71(1-2), 112-117 (2002-08-24)
A remote query magnetoelastic pH sensor comprised of a magnetoelastic thick-film coated with a mass-changing pH-responsive polymer is described. In response to a magnetic query field the magnetoelastic sensor mechanically vibrates at a characteristic frequency that is inversely dependent upon
Crosslinked cinnamoyloxyethyl methacrylate and isooctyl acrylate copolymers as oil sorbers.
Atta A, et al.
Polymer International, 54(7), 1088-1096 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service