Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

436747

Supelco

Amberjet® 4200 Ion Exchange Resin

chloride form, 20-30 mesh

Synonym(s):

Poly(styrene-co-divinylbenzene), quaternary amine functionality, chloride form

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41121813
PubChem Substance ID:

Product Name

Amberjet® 4200 chloride form, chloride form

form

beads

parameter

60 °C Cl- form max. temp.

technique(s)

LPLC: suitable

impurities

50%

matrix

styrene-divinylbenzene (gel)

matrix active group

quaternary ammonium functional group

particle size

600-800 μm

capacity

≥1.30 meq/mL by wetted bed volume

separation technique

anion exchange

SMILES string

[Cl-].C=Cc1ccccc1.C=Cc2ccc(C=C)cc2.C[N+](C)(C)c3ccc(C=C)cc3

Looking for similar products? Visit Product Comparison Guide

General description

Amberjet 4200 chloride form is a strongly basic polystyrene gel resin.[1] It is suitable for removing hexafluoroarsenate from water.[2]

Application

Amberjet 4200 chloride form may be used in separation medium for TLC, paper chromatography and LPLC.
Strongly basic (type I) anion exchange resin for water demineralization; useful for removal of iron and zinc from hydrochloric acid, and recovery of silver from photographic effluents.

Legal Information

Amberjet is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sorption of Cd (II), Co (II), and Zn (II) Complexes with MGDA on Anion Exchange Resins: A Study of the Influence of Various Parameters.
Jachula, Justyna, Dorota Kolodynska, and Zbigniew Hubicki.
Separation Science and Technology, 48, 1801-1809 (2013)
Birgit Daus et al.
Chemosphere, 68(2), 253-258 (2007-02-24)
Arsenic predominantly occurs in natural ground and surface waters as arsenate and arsenite. Other arsenic species can also be present in anthropogenically influenced waters. By means of a newly-developed speciation technique an arsenic compound was identified as hexafluoroarsenate at high

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service