Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

43667

Sigma-Aldrich

4-(Hydroxymethyl)-D-phenylalanine

≥97.0% (HPLC)

Synonym(s):

(R)-2-Amino-3-[4-(hydroxymethyl)phenyl]propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13NO3
CAS Number:
Molecular Weight:
195.22
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

≥97.0% (HPLC)

application(s)

peptide synthesis

SMILES string

N[C@H](Cc1ccc(CO)cc1)C(O)=O

InChI

1S/C10H13NO3/c11-9(10(13)14)5-7-1-3-8(6-12)4-2-7/h1-4,9,12H,5-6,11H2,(H,13,14)/t9-/m1/s1

InChI key

OXNUZCWFCJRJSU-SECBINFHSA-N

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Lempicka et al.
The journal of peptide research : official journal of the American Peptide Society, 53(5), 554-559 (1999-07-29)
This study describes the synthesis and some pharmacological properties of three new analogs of arginine vasopressin (AVP) substituted in position 3 with (R)-alpha-hydroxymethylphenylalanine ([R]-HmPhe). All new peptides were tested for vasopressor and antidiuretic as well as uterotonic activity. None of
M H Kim et al.
International journal of peptide and protein research, 44(5), 457-465 (1994-11-01)
A convenient enantioselective synthesis of p-hydroxymethyl-L-phenylalanine was developed which produces a 4/1 ratio of L/D enantiomers resulting from a chiral phase-transfer-catalyzed alkylation. This amino acid was coupled into the p56(1)ck tyrosine kinase substrate Ac-Leu-Pro-Tyr-Ala-NHCH3 as a replacement for Tyr and
A Olma et al.
Acta biochimica Polonica, 48(4), 1121-1124 (2002-05-09)
To evaluate the role of aromatic amino-acids residues, four analogues of the mu-selective opioid peptide agonist DALDA (H-Tyr-D-Arg-Phe-Lys-NH2) containing the amphiphilic, a,a-disubstituted amino acid (R)- or (S)-alpha-hydroxymethyltyrosine (HmTyr) in position 1 and (R)- or (S)-alpha-hydroxymethylphenylalanine (HmPhe) in position 3 of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service