Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

435252

Sigma-Aldrich

Poly(styrene-co-maleic acid), partial isobutyl/methyl mixed ester

average Mv ~180,000, powder

Synonym(s):

maleated copolymer, maleated polymer, maleic copolymer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH2CH(C6H5)]x[CH(CO2R)CH(CO2R)]y, R = H or CH3 or-CH2CH(CH3)2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

form

powder

mol wt

average Mv ~180,000

viscosity

15-35 cP, 10 %(25 °C, Brookfield, spindle #1) (60 rpm, pH 8.3)(lit.)

acid number

~185 mg KOH/g

transition temp

Tg 140-150 °C
flow temperature 230-235 °C

solubility

aqueous base, alcohols, ketones and ethyl acetate: soluble

density

1.15 g/mL at 25 °C (lit.)

SMILES string

OC(=O)\C=C\C(O)=O.C=Cc1ccccc1.COC(=O)\C=C\C(=O)OC.CC(C)COC(=O)\C=C\C(=O)OCC(C)C

Looking for similar products? Visit Product Comparison Guide

General description

1:<1 mole ratio of styrene:maleic anhydride

Application

Dispersant, emulsifier, binder, sizing agent and viscosity modifier.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service