Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

434795

Sigma-Aldrich

2-Benzyloxy-3-methoxybenzaldehyde

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH2OC6H3(OCH3)CHO
CAS Number:
Molecular Weight:
242.27
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

98%

form

solid

mp

44-46 °C (lit.)

functional group

aldehyde
phenyl

SMILES string

COc1cccc(C=O)c1OCc2ccccc2

InChI

1S/C15H14O3/c1-17-14-9-5-8-13(10-16)15(14)18-11-12-6-3-2-4-7-12/h2-10H,11H2,1H3

InChI key

KXBGOVZWCRLLOR-UHFFFAOYSA-N

General description

2-Benzyloxy-3-methoxybenzaldehyde (benzyl-o-vanillin) can be prepared from the reaction of o-vanillin with benzyl bromide in acetone (solvent) and K2CO3 (base) in the presence of tetra-n-butylammonium iodide (catalyst).[1] The dihedral angle between the two benzene rings in 2-benzyloxy-3-methoxybenzaldehyde crystal is 23.33 (6)°. Crystal structure suggests that all O atoms are involved in intra and intermolecular C-HO interactions.[2] It has been reported as key reagent for the synthesis of new anticancer drugs.[1] It is an important pharmacophore in drug discovery. It exhibits anti-proliferative activity in HL60 leukemia cancer cells.[1][3]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zena A Al-Mudaris et al.
PloS one, 8(11), e80983-e80983 (2013-11-22)
Benzyl-o-vanillin and benzimidazole nucleus serve as important pharmacophore in drug discovery. The benzyl vanillin (2-(benzyloxy)-3-methoxybenzaldehyde) compound shows anti-proliferative activity in HL60 leukemia cancer cells and can effect cell cycle progression at G2/M phase. Its apoptosis activity was due to disruption
2-Benzyloxy-3-methoxybenzaldehyde (benzyl-o-vanillin).
Al-Douh MH, et al.
Acta Crystallographica Section E, Structure Reports Online, 62(11), 4768-4770 (2006)
1D and 2D NMR Studies of Benzyl o-Vanillin.
Al-Douh MH, et al.
Indonesian Journal of Chemistry, 8(3), 411-417 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service