434523
Acetone-d6
≥99.9 atom % D, contains 0.1 % (v/v) TMS
Synonym(s):
Hexadeuteroacetone
About This Item
Recommended Products
vapor density
2 (vs air)
vapor pressure
14.39 psi ( 55 °C)
184 mmHg ( 20 °C)
3.67 psi ( 20 °C)
isotopic purity
≥99.9 atom % D
assay
≥99% (CP)
form
liquid
contains
0.1 % (v/v) TMS
expl. lim.
13.2 %
packaging
ampule of 10 g
impurities
≤0.0500% water
water
refractive index
n20/D 1.355 (lit.)
bp
55.5 °C (lit.)
mp
−93.8 °C (lit.)
density
0.872 g/mL at 25 °C (lit.)
mass shift
M+6
SMILES string
[2H]C([2H])([2H])C(=O)C([2H])([2H])[2H]
InChI
1S/C3H6O/c1-3(2)4/h1-2H3/i1D3,2D3
InChI key
CSCPPACGZOOCGX-WFGJKAKNSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
accessory
signalword
Danger
hcodes
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk_germany
WGK 1
flash_point_f
-2.2 °F - closed cup
flash_point_c
-19 °C - closed cup
ppe
Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service