Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

434477

Sigma-Aldrich

Poly(vinyl acetate-co-butyl maleate-co-isobornyl acrylate) solution

50 wt. % in ethanol

Synonym(s):

2-Butenedioic acid (Z )-, dibutyl ester, polymer with ethenyl acetate and, Butyl maleate-isobornyl acrylate-vinyl acetate copolymer, Vinyl acetate-butyl maleate-isobornyl acrylate copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

form

viscous liquid

composition

butyl maleate, ~57 wt. %

concentration

50 wt. % in ethanol

refractive index

n20/D 1.4158

relative viscosity

1.31 v, 1 % (w/v) in ethanol(lit.)

bp

78 °C

acid number

185 mg KOH/g(dry basis)

solubility

soluble (Water solubility is controlled by the degree of polymerization.)

density

0.957 g/mL at 25 °C

Application

Additive to hair and skin care products.

pictograms

FlameExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

48.2 °F - closed cup

flash_point_c

9 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service