Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

434469

Sigma-Aldrich

Poly(1-vinylpyrrolidone-co-2-dimethylaminoethyl methacrylate) solution

average Mw ~1,000,000 by GPC, ≥19 wt. % in H2O

Synonym(s):

2-(Dimethylamino)ethyl methacrylate-N -vinyl pyrrolidone copolymer, Dimethylaminoethyl methacrylate-N -vinylpyrrolidinone copolymer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352300
NACRES:
NA.23

form

liquid

mol wt

average Mw ~1,000,000 by GPC

concentration

≥19 wt. % in H2O

refractive index

n20/D 1.372

pH

6-8

viscosity

20,000 cP(25 °C, Brookfield)(lit.)

solubility

alcohols and water: soluble

density

1.047 g/mL at 25 °C

InChI

1S/C8H15NO2.C6H9NO/c1-7(2)8(10)11-6-5-9(3)4;1-2-7-5-3-4-6(7)8/h1,5-6H2,2-4H3;2H,1,3-5H2

Inchi Key

SJQISFXXYCYMIK-UHFFFAOYSA-N

Application

Additive to hair and skin care products.

Other Notes

Film former.

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Self-assembled monolayers (SAMs) have diverse applications; article compares benefits of alkylthiolates on gold SAM systems.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service