Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

431702

Sigma-Aldrich

N,N′,N′′-Trimethylbis(hexamethylene)triamine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3N[(CH2)6NHCH3]2
CAS Number:
Molecular Weight:
257.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

refractive index

n20/D 1.462 (lit.)

density

0.857 g/mL at 25 °C (lit.)

SMILES string

CNCCCCCCN(C)CCCCCCNC

InChI

1S/C15H35N3/c1-16-12-8-4-6-10-14-18(3)15-11-7-5-9-13-17-2/h16-17H,4-15H2,1-3H3

InChI key

XFKPZJBVIVCMKS-UHFFFAOYSA-N

General description

N,N′,N′′-Trimethylbis(hexamethylene)triamine (N,N′-dimethyl-N-[6-(methylamino)hexyl]-1,6-hexanediamine) is a polyamine.

Application

N,N′,N′′-Trimethylbis(hexamethylene)triamine is suitable amine employed to investigate the stability of complexes formed by dicarboxylic ligands with open-chain polyammonium cations by potentiometry.[1]

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

235.0 °F - closed cup

flash_point_c

112.8 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stability-Charge and Stability-Structure relationships in the binding of dicarboxylic ligands by open-chain polyammonium cations.
Cascio S, et al.
Journal of Chemical and Engineering Data, 45(5), 717-723 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service