431532
Ammonium persulfate
ReagentPlus®, ≥99.99%
Synonym(s):
AP, APS, Ammonium peroxodisulfate, Ammonium peroxydisulfate, PER
About This Item
Recommended Products
vapor density
7.9 (vs air)
product line
ReagentPlus®
assay
≥99.99%
impurities
≤0.005% insolubles
≤0.04 meq/g Titr. Acid
<100 ppm total metallic impurities
ign. residue
≤0.05%
anion traces
Cl- and ClO4-: ≤0.001%
cation traces
Fe: ≤0.001%
Mn: ≤0.5 ppm
heavy metals: ≤0.005%
storage temp.
2-8°C
SMILES string
N.N.OS(=O)(=O)OOS(O)(=O)=O
InChI
1S/2H3N.H2O8S2/c;;1-9(2,3)7-8-10(4,5)6/h2*1H3;(H,1,2,3)(H,4,5,6)
InChI key
ROOXNKNUYICQNP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Features and Benefits
Legal Information
replaced by
signalword
Danger
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 3 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
target_organs
Respiratory system
Storage Class
5.1B - Oxidizing hazardous materials
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service