Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

431281

Sigma-Aldrich

Brij® O10

average Mn ~709

Synonym(s):

Brij 97, C18-1E10, Polyoxyethylene (10) oleyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18H35(OCH2CH2)nOH, n~10
CAS Number:
MDL number:
UNSPSC Code:
12162002

mol wt

average Mn ~709

density

1 g/mL at 25 °C (lit.)

HLB

12

InChI

1S/C20H40O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-19-22-20-18-21/h9-10,21H,2-8,11-20H2,1H3/b10-9-

InChI key

KWVPFECTOKLOBL-KTKRTIGZSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

Brij is a registered trademark of Croda International PLC

pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Chronic 2 - Skin Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

>464.0 °F - Equilibrium method

flash_point_c

> 240 °C - Equilibrium method

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carles Gil et al.
Biochemical and biophysical research communications, 348(4), 1334-1342 (2006-08-22)
Although the high presence of cholesterol in nerve terminals is well documented, specific roles of this lipid in transmitter release have remained elusive. Since cholesterol is a highly enriched component in the membrane microdomains known as lipid rafts, it is
Sergey Filippov et al.
Langmuir : the ACS journal of surfaces and colloids, 24(17), 9295-9301 (2008-08-09)
In this work we report a new type of pH-responsive micelle-like nanoparticle. Reversible nanoscale structures are formed in solutions of a pH-sensitive hydrophobic polyelectrolyte, poly( N-methacryloyl- l-valine) or poly( N-methacryloyl- l-phenylalanine), and nonionic surfactant (Brij 98) in the presence of
Jia-You Fang et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 67(1), 67-75 (2007-02-27)
Acoustically active lipospheres (AALs) were prepared using perfluorocarbons and coconut oil as the cores of inner phase. These AALs were stabilized using coconut oil and phospholipid coatings. A lipophilic antioxidant, resveratrol, was the model drug loaded into the AALs. AALs
Yash Kapoor et al.
Journal of colloid and interface science, 322(2), 624-633 (2008-03-25)
Surfactants are commonly incorporated into hydrogels to increase solute loading and attenuate the release rates. In this paper we focus on understanding and modeling the mechanisms of both surfactant and drug transport in hydrogels. Specifically, we focus on Brij 98
Jessica Phelps et al.
Colloids and surfaces. B, Biointerfaces, 87(2), 391-398 (2011-06-28)
In this study, fluid precursor formulations for subcutaneous injection and in situ formation of hexagonal phase gels upon water absorption were developed as a strategy to sustain the release of naltrexone, a drug used for treatment of drug addiction. Precursor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service