Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

430528

Sigma-Aldrich

Poly(1-vinylpyrrolidone)-graft-(1-triacontene)

flakes

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

flakes

impurities

2-3% Nitrogen

relative viscosity

1.2-1.5, 5 wt. % in toluene(30 °C, Cannon-Fenske #100)(lit.)

bromine number

10

transition temp

Tg 46 °C

General description

Film former with surface activity, water resistance and barrier properties.

Application

Pigment dispersant and wetting agent, substantive to skin.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service