Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

430250

Sigma-Aldrich

Trimethylolethane triglycidyl ether

technical grade

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24O6
CAS Number:
Molecular Weight:
288.34
UNSPSC Code:
12352001
Pricing and availability is not currently available.

grade

technical grade

eq. wt.

155-175 (perchloric acid method)

impurities

≤10 ppm free epichlorohydrin

viscosity

200-330 cSt(25 °C, Ubbelohde)(lit.)

InChI

1S/C5H12O3.C3H5ClO/c1-5(2-6,3-7)4-8;4-1-3-2-5-3/h6-8H,2-4H2,1H3;3H,1-2H2

InChI key

OAFLYVWDZVWXCD-UHFFFAOYSA-N

Application

Additive in fast-setting adhesives, low temperature curing coatings and hard, abrasion resistant castings and decoupage systems.

Features and Benefits

Reduces viscosity but retains high epoxide level, imparts hardness and toughness to epoxide polymers. Improves solubility/compatibility characteristics of highly aromatic epoxy systems.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service