Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

429872

Sigma-Aldrich

2,3-Thiophenedicarboxaldehyde

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4O2S
CAS Number:
Molecular Weight:
140.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

97%

form

solid

mp

76-78 °C (lit.)

SMILES string

O=Cc1ccsc1C=O

InChI

1S/C6H4O2S/c7-3-5-1-2-9-6(5)4-8/h1-4H

InChI key

WSEJZRIZDQWMKQ-UHFFFAOYSA-N

General description

2,3-Thiophenedicarboxaldehyde undergoes aldol reaction with (4aS,9R,9aR,10S)-11-(propan-2-ylidene)-2,3,4a,9,9a,10-hexahydro-9,10-methanoanthracene-1,4-dione under oxygen-free conditions affords (5aS,6S,11R,11aR)-14-(propan-2-ylidene)-5a,6,11,11a-tetrahydro-6,11-methanotetraceno[3,2-b]thiophene-5,12-dione.[1]

Application

2,3-Thiophenedicarboxaldehyde may be employed for the synthesis of TIPSE (triisopropylsilylethynyl)-substituted dibromoanthradithiophene (TIPSEBr2- AntDT).[2]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Characterization of Pentacene- and Anthradithiophene- Fluorene Conjugated Copolymers Synthesized by Suzuki Reactions.
Okamoto T, et al.
Macromolecules, 41(19), 6977-6980 (2008)
Solution-processed organic micro crystal transistor based on tetraceno [2, 3-b] thiophene from a monoketone precursor.
Watanabe M, et al.
Journal of Materials Chemistry, 21(30), 11317-11322 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service