Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

428272

Sigma-Aldrich

Poly(trimellitic anhydride chloride-co-4,4′-methylenedianiline)

powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

powder

refractive index

n20/D 1.656 (film)(lit.)

viscosity

800 cP

mp

>300 °C (lit.)

acid number

80 mg KOH/g

transition temp

softening point 272 °C (Film)

density

1.11 g/mL at 25 °C (lit.)

Application

Used in conformal coatings, passivation agent, insulating varnish, LCD alignments, and corrosion resistant coatings.

Physical properties

Reactive, low molecular weight, soluble polymer. Undergoes imide formation at 100-150°C. Continued heating to 150°C removes residual solvent.

pictograms

Health hazardExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Repr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P2 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service