427853
Polypropylene
Isotactic, average Mw ~580,000, average Mn ~166,000
About This Item
melt index
0.5 g/10 min (230°C/2.16kg)
mol wt
average Mn ~166,000
average Mw ~580,000
hardness
95 (Rockwell R, ASTM D 785-A)
transition temp
Tm (DSC) 160-165 °C (onset)
density
0.9 g/mL at 25 °C (lit.)
SMILES string
CC=C
InChI
1S/C22H42O3/c1-2-3-4-5-11-14-17-20-21(25-20)18-15-12-9-7-6-8-10-13-16-19-22(23)24/h20-21H,2-19H2,1H3,(H,23,24)/t20-,21+/m1/s1
InChI key
NSYDMBURIUSUDH-RTWAWAEBSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Features and Benefits
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service