Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

427276

Sigma-Aldrich

Poly[1,4-benzenedicarbonyl-alt-bis(4-phenoxyphenyl)methanone]

melt index 200 g/10 min (400°C/10kg, ISO 1311)

Synonym(s):

PAEK (polyaryletherketone)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H4OC6H4COC6H4OC6H4COC6H4CO)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

pellets

melt index

200 g/10 min (400°C/10kg, ISO 1311)

viscosity

100, 5 g/L in concentrated sulfuric acid(25 °C)(lit.)

mp

364 °C

transition temp

Tg 158 °C (DSC)
softening point >240 °C (Vicat)

density

1.3 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Application

Functional parts for household appliances, automotive and precision engineered components and general applications requiring high temperature and chemical resistance.

Features and Benefits

Favorable tribological and dielectric properties. Extreme resistance to hydrolysis and little tendency to stress crack.

Physical form

Semicrystalline thermoplastic

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service