Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

427195

Sigma-Aldrich

Poly(ethylene-co-tetrafluoroethylene)

melt index 6 g/10 min (279°C/49 newton), pellets

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH2CH2)x(CF2CF2)y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

description

coefficient of friction 0.53 Dynamic, p=110 lbs,
velocity=0.08-0.35 cm/sec

form

pellets

melt index

6 g/10 min (279°C/49 newton)

hardness

50-58 (Rockwell R, ASTM D 785)

transition temp

brittleness temperature -125 °C (ASTM D 746)
Tm 250-270 °C (ASTM D 3418)

density

1.73 g/mL at 25 °C (lit.)

SMILES string

C=C.F\C(F)=C(\F)F

InChI

1S/C2F4.C2H4/c3-1(4)2(5)6;1-2/h;1-2H2

InChI key

QHSJIZLJUFMIFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Compression and transfer molding for pipes, sheets and cable jackets.

Features and Benefits

Mechanical properties are superior to perfluorinated polymers like PTFE and PFA. Excellent performance over a wide temperature range and good electrical insulating properties over a wide temperature and frequency range.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M L Raimondi et al.
Journal of biomedical materials research, 29(1), 59-63 (1995-01-01)
The release behavior of typical commercial tissue expanders has been examined by carrying out two kinds of experiments: 1) Determination of chemical nature and its modification after in vivo use by X-ray photoelectron spectroscopy for chemical analysis. This study has
T Paulos et al.
Risk analysis : an official publication of the Society for Risk Analysis, 18(4), 471-510 (1998-10-17)
This paper investigates electrical overheating events aboard a habitable spacecraft. The wire insulation involved in these failures plays a major role in the entire event scenario from threat development to detection and damage assessment. Ideally, if models of wire overheating
Kristin J Pawlowski et al.
Frontiers in bioscience : a journal and virtual library, 9, 1412-1421 (2004-02-24)
Pioneers in the field of small diameter graft development sought to promote graft endothelialization and, thereby, increase patency by transplanting a varying degree of autologous endothelial cells onto vascular grafts prior to implantation. This process has become known as endothelial
Anthony G Gegauff et al.
Dental materials : official publication of the Academy of Dental Materials, 18(6), 479-485 (2002-07-06)
This pilot study was conducted to explore selected commercially-available thermoplastic composites that potentially had physical properties superior to currently available dental systems for restoring esthetic posterior crowns. Polyurethane, polycarbonate, and poly(ethylene/tetrafluoroethylene) (ETFE) composites and unfilled polyurethane specimens were injection molded
H Kaplan et al.
The Biochemical journal, 217(1), 135-143 (1984-01-01)
A modification to the competitive labelling procedure of Duggleby and Kaplan [(1975) Biochemistry 14, 5168-5175] was used to study the reactivity of the N-termini, lysine, histidine and tyrosine groups of insulin over the concentration range 1 X 10(-3)-1 X 10(-7)M.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service