Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

426598

Sigma-Aldrich

Poly(1,4-butanediol)bis(4-aminobenzoate)

average Mn ~470

Synonym(s):

Polytetramethylene glycol bis(4-aminobenzoate), Polytetramethylene oxide di-p -aminobenzoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CO2(C4H8O)nC(O)C6H4NH2
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

mol wt

average Mn ~470
degree of polymerization 3-4

viscosity

300 cP(85 °C)(lit.)

mp

56 °C

density

1.07 g/mL at 25 °C

SMILES string

OCCCCO.Nc1ccc(cc1)C(O)=O

InChI

1S/C18H20N2O4/c19-15-7-3-13(4-8-15)17(21)23-11-1-2-12-24-18(22)14-5-9-16(20)10-6-14/h3-10H,1-2,11-12,19-20H2

InChI key

UCHUCWHZVSIPQI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

application

Room temperature vulcanizates (RTV) or thermally cured molded parts. Encapsulation of temperature-sensitive electronic components.

Features and Benefits

Reacts with diisocyanate prepolymers at room or elevated temperatures to give polyurea polymers.

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

235.4 °F - closed cup

flash_point_c

113 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service