425753
4-Nitrophenol
reagent grade, 98%
Synonym(s):
p-Nitrophenol
About This Item
Recommended Products
grade
reagent grade
vapor pressure
0.6 mmHg ( 120 °C)
assay
98%
form
solid
autoignition temp.
541 °F
bp
279 °C (lit.)
mp
110-115 °C (lit.)
SMILES string
O=N(C1=CC=C(O)C=C1)=O
InChI
1S/C6H5NO3/c8-6-3-1-5(2-4-6)7(9)10/h1-4,8H
InChI key
BTJIUGUIPKRLHP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
replaced by
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - STOT RE 2 Oral
target_organs
Kidney,Liver
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk_germany
WGK 2
flash_point_f
336.2 °F
flash_point_c
169 °C
ppe
dust mask type N95 (US), Eyeshields, Faceshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service