Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

423394

Sigma-Aldrich

(R)-(−)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride

99%, ChiraSelect

Synonym(s):

(R)-(−)-MTPA-Cl, Mosher’s acid chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(OCH3)(CF3)COCl
CAS Number:
Molecular Weight:
252.62
Beilstein/REAXYS Number:
3591563
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

product line

ChiraSelect

assay

99%

optical purity

ee: ≥99% (GLC)

refractive index

n20/D 1.47 (lit.)

bp

213-214 °C (lit.)

density

1.35 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

CO[C@](C(Cl)=O)(c1ccccc1)C(F)(F)F

InChI

1S/C10H8ClF3O2/c1-16-9(8(11)15,10(12,13)14)7-5-3-2-4-6-7/h2-6H,1H3/t9-/m0/s1

InChI key

PAORVUMOXXAMPL-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Miller et al.
Journal of chromatography, 307(2), 335-342 (1984-05-11)
High-performance liquid chromatography (HPLC) was employed for resolution of enantiomers of chiral ring-substituted 1-phenyl-2- aminopropanes (amphetamines) and 1-phenylethylamine following derivatization with four chiral reagents: (R)-(+)-1-phenylethyl isocyanate ( PEIC ), (-)-alpha-methoxy-alpha-(trifluoromethyl)phenylacetyl chloride ( MTPA X Cl), 2,3,4,6-tetra-O-acetyl-beta-D-glucopyranosyl isothiocyanate ( GITC )

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service