Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

422045

Sigma-Aldrich

DBE-9

Synonym(s):

Dibasic ester mixture, mixture of dimethyl glutarate and dimethyl succinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3O2C(CH2)nCO2CH3 (n=2,3)
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

vapor pressure

0.2 mmHg ( 20 °C)

form

liquid

autoignition temp.

698 °F

expl. lim.

8 %

refractive index

n20/D 1.422 (lit.)

bp

196-215 °C (lit.)

density

1.099 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCC(=O)OC.COC(=O)CCCC(=O)OC

InChI

1S/C7H12O4.C6H10O4/c1-10-6(8)4-3-5-7(9)11-2;1-9-5(7)3-4-6(8)10-2/h3-5H2,1-2H3;3-4H2,1-2H3

InChI key

BNMYXGKEMMVHOX-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

201.2 °F - closed cup

flash_point_c

94 °C - closed cup

ppe

Eyeshields, Gloves, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service