Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

421936

Sigma-Aldrich

(R)-(+)-α-Methylbenzylamine

≥99%, ChiraSelect

Synonym(s):

(R)-(+)-1-Phenylethylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(CH3)NH2
CAS Number:
Molecular Weight:
121.18
Beilstein/REAXYS Number:
2410916
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Pricing and availability is not currently available.

vapor pressure

0.5 mmHg ( 20 °C)

product line

ChiraSelect

assay

≥99%

optical purity

ee: ≥99% (GLC)

refractive index

n20/D 1.526 (lit.)

bp

187-189 °C (lit.)

density

0.952 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C[C@@H](N)c1ccccc1

InChI

1S/C8H11N/c1-7(9)8-5-3-2-4-6-8/h2-7H,9H2,1H3/t7-/m1/s1

InChI key

RQEUFEKYXDPUSK-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiraSelect is a trademark of Sigma-Aldrich Co. LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karim Engelmark Cassimjee et al.
Organic & biomolecular chemistry, 10(28), 5466-5470 (2012-06-13)
For biocatalytic production of pharmaceutically important chiral amines the ω-transaminase enzymes have proven useful. Engineering of these enzymes has to some extent been accomplished by rational design, but mostly by directed evolution. By use of a homology model a key
Sachin R Chaudhari et al.
Organic & biomolecular chemistry, 10(31), 6410-6419 (2012-06-28)
The three-component chiral derivatization protocols have been developed for (1)H, (13)C and (19)F NMR spectroscopic discrimination of chiral diacids by their coordination and self-assembly with optically active (R)-α-methylbenzylamine and 2-formylphenylboronic acid or 3-fluoro-2-formylmethylboronic acid. These protocols yield a mixture of
Hyungdon Yun et al.
Bioscience, biotechnology, and biochemistry, 72(11), 3030-3033 (2008-11-11)
To produce (S)-alpha-methylbenzylamine (MBA) from acetophenone, recombinant Escherichia coli co-expressing omega-transaminase and acetolactate synthase was used as a whole-cell biocatalyst. The solvent-bridge reaction system increased the yield of the whole-cell reaction by 2.5-fold, and the inhibitory (S)-alpha-MBA produced in the
Shao-Hua Huang et al.
Magnetic resonance in chemistry : MRC, 47(5), 423-427 (2009-02-19)
Two types of chiral analytes, the urea and amide derivatives of alpha-phenylethylamine, were prepared. The effect of inter-molecular hydrogen-bonding interaction on self-discrimination of the enantiomers of analytes has been investigated using high-resolution (1)H NMR. It was found that the urea
Manuel Algarra et al.
Journal of separation science, 33(2), 143-154 (2009-12-31)
Free fatty acids were derivatized as amides (DFFA) by reaction with (R)-(+)-1-phenylethylamine, using a simple, fast and robust reaction scheme. A HPLC method with diode array and ESI MS detection was developed for the analysis of the derivatized substances. Six

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service