Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420522

Sigma-Aldrich

ALKANOL® 189-S surfactant

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002

refractive index

n20/D 1.415 (lit.)

bp

100 °C (lit.)

density

1.06 g/mL at 25 °C (lit.)

Legal Information

DuPont Product
ALKANOL is a registered trademark of E. I. du Pont de Nemours and Company

signalword

Danger

Hazard Classifications

Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Skin Corr. 1A - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

69.8 °F

flash_point_c

21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service