Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

420174

Sigma-Aldrich

cis-3-Hydroxy-DL-proline

95%

Synonym(s):

(±)-cis-3-Hydroxypyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO3
CAS Number:
Molecular Weight:
131.13
Beilstein/REAXYS Number:
4130229
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

assay

95%

reaction suitability

reaction type: solution phase peptide synthesis

mp

232 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

O[C@@H]1CCN[C@@H]1C(O)=O

InChI

1S/C5H9NO3/c7-3-1-2-6-4(3)5(8)9/h3-4,6-7H,1-2H2,(H,8,9)/t3-,4+/m1/s1

InChI key

BJBUEDPLEOHJGE-DMTCNVIQSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mary Ann Weis et al.
The Journal of biological chemistry, 285(4), 2580-2590 (2009-11-27)
Collagen triple helices are stabilized by 4-hydroxyproline residues. No function is known for the much less common 3-hydroxyproline (3Hyp), although genetic defects inhibiting its formation cause recessive osteogenesis imperfecta. To help understand the pathogenesis, we used mass spectrometry to identify
Sanjay Morzaria et al.
The Journal of otolaryngology, 34(5), 297-303 (2005-09-27)
To develop an evidence-based algorithm for determining the etiology of bilateral sensorineural hearing loss (SNHL) in a child. The frequency of different etiologies was previously determined. A systematic review of the literature for articles published between 1940 and January 2003
David M Hudson et al.
PloS one, 6(5), e19336-e19336 (2011-05-12)
Recessive mutations that prevent 3-hydroxyproline formation in type I collagen have been shown to cause forms of osteogenesis imperfecta. In mammals, all A-clade collagen chains with a GPP sequence at the A1 site (P986), except α1(III), have 3Hyp at residue
B Lubec et al.
The Journal of pediatrics, 123(5), 748-751 (1993-11-01)
Because disturbed conformation of connective tissue proteins can be accompanied by increased racemization (i.e., an increased ratio of dextrorotatory (D) to levorotatory (L) amino acid molecules), we studied by high-performance liquid chromatography the renal excretion of the D-form of the
B Bartosch et al.
Archives of disease in childhood, 66(2), 248-251 (1991-02-01)
Alport's syndrome is characterised by morphological and structural changes of the renal basement membranes. As the hydroxyproline content of isolated glomerular basement membranes is reduced in patients with Alport's syndrome, it is possible that the renal excretion of 3-hydroxproline (3-OHP)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service