Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

419540

Sigma-Aldrich

1-Propenylmagnesium bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=CHMgBr
CAS Number:
Molecular Weight:
145.28
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

bp

65 °C

density

0.95 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

CC=C[Mg]Br

InChI

1S/C3H5.BrH.Mg/c1-3-2;;/h1,3H,2H3;1H;/q;;+1/p-1

InChI key

HHNLJRLNDSEYOX-UHFFFAOYSA-M

Application

1-Propenylmagnesium bromide solution can be used in the preparation of:
  • Titanocene alkenylidene intermediates in situ from alkenyltitanocene precursors, which then reacts with carbonyl compounds to form the corresponding allenes.[1]
  • 1-(4-ethoxy-phenyl)-3-methyl-hex-4-en-1-one from methyl 4-ethoxybenzoate.[2]
  • 2-(propen-1-yl)chroman-4-ones from chromones.[3]

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

target_organs

Central nervous system, Respiratory system

supp_hazards

Storage Class

4.3 - Hazardous materials which set free flammable gases upon contact with water

wgk_germany

WGK 3

flash_point_f

-4.0 °F - closed cup

flash_point_c

-20 °C - closed cup

ppe

Faceshields, Gloves, Goggles


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allenation of carbonyl compounds with alkenyltitanocene derivatives.
Petasis N A and Hu Y H
The Journal of Organic Chemistry, 62(4), 782-783 (1997)
Synthesis and antimicrobial activity of 2-alkenylchroman-4-ones, 2-alkenylthiochroman-4-ones and 2-alkenylquinol-4-ones
Hoettecke N, et al.
Bioorganic & Medicinal Chemistry, 16(24), 10319-10325 (2008)
Efficient synthesis of apricoxib, CS-706, a selective cyclooxygenase-2 inhibitor, and evaluation of inhibition of prostaglandin E2 production in inflammatory breast cancer cells.
Mandal P K, et al.
Bioorganic & Medicinal Chemistry Letters, 21(20), 6071-6073 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service