Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

419176

Sigma-Aldrich

4-Aminomethyl-1,8-octanediamine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H23N3
CAS Number:
Molecular Weight:
173.30
Beilstein/REAXYS Number:
3029083
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Pricing and availability is not currently available.

assay

95%

form

solid

SMILES string

NCCCCC(CN)CCCN

InChI

1S/C9H23N3/c10-6-2-1-4-9(8-12)5-3-7-11/h9H,1-8,10-12H2

InChI key

HMJBXEZHJUYJQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service