Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

418595

Sigma-Aldrich

Poly(melamine-co-formaldehyde) isobutylated, solution

average Mn ~1,620, 50 wt. % in isobutanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

liquid

mol wt

average Mn ~1,620

concentration

50 wt. % in isobutanol

refractive index

n20/D 1.468

bp

108 °C

density

1 g/mL at 25 °C

InChI

1S/C3H6N6.CH2O/c4-1-7-2(5)9-3(6)8-1;1-2/h(H6,4,5,6,7,8,9);1H2

Inchi Key

IVJISJACKSSFGE-UHFFFAOYSA-N

Application

Cross-linking agent.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Carc. 1B - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

target_organs

Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

77.0 °F

flash_point_c

25 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service