Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

418439

Sigma-Aldrich

Poly(vinyl butyral-co-vinyl alcohol-co-vinyl acetate)

average Mw 70,000-100,000, powder

Synonym(s):

Poly(vinyl butyral-vinyl alcohol-vinyl acetate)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002

form

powder

mol wt

average Mw 70,000-100,000

composition

acetate, 0-1.5 wt. %
hydroxyl, 18-20 wt. %
vinyl butyral, 80 wt. %

viscosity

6.0-9.0 cSt, 6.0 wt. % in methanol(20 °C, Ostwald)(lit.)

transition temp

Tg 72-78 °C

solubility

alcohols, glycol ethers and dioxane: soluble

density

1.083 g/mL at 25 °C (lit.)

InChI

1S/C8H14O2.C4H6O2.C2H4O/c1-4-7-8(9-5-2)10-6-3;1-3-6-4(2)5;1-2-3/h5-6,8H,2-4,7H2,1H3;3H,1H2,2H3;2-3H,1H2

InChI key

RNNFCYZWIQZWQV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ceramic binder, dry toners, wood coatings, composite fiber binders, structural adhesives and interlayer in safety glass.

Features and Benefits

Good binding efficiency, optical clarity and adhesion to a variety of surfaces.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service