Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

417025

Sigma-Aldrich

Styryl 9M

Dye content ~98 %

Synonym(s):

2-([3-(2-[4-(Dimethylamino)phenyl]ethenyl)-5,5-dimethyl-2-cyclohexen-1-ylidene]methyl)-3-methylbenzothiazolium perchlorate, 2-[[3-[2-[4-(Dimethylamino)phenyl]ethenyl]-5,5-dimethyl-2-cyclohexen-1-ylidene]methyl]-3-methylbenzothiazolium perchlorate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31ClN2O4S
CAS Number:
Molecular Weight:
515.06
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

composition

Dye content, ~98%

mp

250 °C (dec.) (lit.)

λmax

584 nm

SMILES string

[O-]Cl(=O)(=O)=O.CN(C)c1ccc(\C=C\C2=C\C(CC(C)(C)C2)=C/c3sc4ccccc4[n+]3C)cc1

InChI

1S/C27H31N2S.ClHO4/c1-27(2)18-21(11-10-20-12-14-23(15-13-20)28(3)4)16-22(19-27)17-26-29(5)24-8-6-7-9-25(24)30-26;2-1(3,4)5/h6-17H,18-19H2,1-5H3;(H,2,3,4,5)/q+1;/p-1

InChI key

ZVJQLQFMYXZEDW-UHFFFAOYSA-M

Application

Suitable as a laser dye

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiangquan Deng et al.
Journal of biophotonics, 13(12), e202000299-e202000299 (2020-10-08)
Indocyanine green (ICG) is a human compatible dye and is ideal for deep-tissue two-photon fluorescence (2PF) microscopy excited at the 1700-nm window in vivo. However, the two-photon excitation and emission properties of this dye remain unknown. Here we demonstrate measurement
Lauren M Barnett et al.
PloS one, 12(2), e0170934-e0170934 (2017-02-10)
The goal of this work is to determine how GCaMP6m's fluorescence is altered in response to Ca2+-binding. Our detailed spectroscopic study reveals the simplest explanation for how GCaMP6m changes fluorescence in response to Ca2+ is with a four-state model, in

Articles

Fluorescence quenching microscopy visualizes 2D materials like graphene and MoS2 rapidly, inexpensively, and with high fidelity.

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service