Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

416053

Sigma-Aldrich

Poly(acrylic acid-co-maleic acid) solution

average Mw 3,000, 50 wt. % in H2O

Synonym(s):

Acrylic acid-maleic acid copolymer, Poly(acrylic acid-maleic acid)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

viscous liquid

mol wt

average Mw 3,000

concentration

50 wt. % in H2O

refractive index

n20/D 1.423

viscosity

100 cP(23 °C)

density

1.23 g/mL at 25 °C

SMILES string

OC(=O)C=C.OC(=O)\C=C\C(O)=O

General description

1:1 mole ratio of AA:MA

pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 3

flash_point_f

212.0 °F - closed cup

flash_point_c

100 °C - closed cup

ppe

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laura Mayne et al.
ACS nano, 12(5), 4844-4852 (2018-05-03)
Aptamer-modified nanomaterials provide a simple, yet powerful sensing platform when combined with resistive pulse sensing technologies. Aptamers adopt a more stable tertiary structure in the presence of a target analyte, which results in a change in charge density and velocity

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service