Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

414611

Sigma-Aldrich

Palladium on strontium carbonate

reduced

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

SMILES string

[Pd].OC(=O)O[Sr]OC(O)=O

InChI

1S/2CH2O3.Pd.Sr/c2*2-1(3)4;;/h2*(H2,2,3,4);;/q;;;+2/p-2

InChI key

OVQPMFIBJZVJSC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for the preparation of a variety of organic molecules[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalysis of Organic Reactions
M. G. Scaros, et al.,
Org. React., 307-314 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service